Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnmtl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183263.5 | NP_899086.2 | 4 | 1409 | ||
GenBank mRNA | AK012200.1 | - | 4 | 1384 | ||
AK014739.1 | - | 4 | 1375 | |||
AK147805.1 | - | 4 | 1390 | |||
BC090975.1 | - | 4 | 1381 | 6595681 |