Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256702.1 | NP_001243631.1 | 11, 12 | 1320 | ||
NM_006537.3 | NP_006528.2 | 12, 13 | 1452 | |||
NR_046341.1 | 13, 14 | 1605 | ||||
NR_046342.1 | 13, 14 | 1568 | ||||
XM_017022763.1 | XP_016878252.1 | 14, 15 | 2451 | |||
XM_017022764.1 | XP_016878253.1 | 13, 14 | 2387 | |||
XM_017022765.1 | XP_016878254.1 | 13, 14 | 1593 | |||
XM_017022766.1 | XP_016878255.1 | 14, 15 | 1636 | |||
XM_017022767.1 | XP_016878256.1 | 15, 16 | 2567 | |||
XM_017022768.1 | XP_016878257.1 | 15, 16 | 2641 | |||
GenBank mRNA | AF073344.1 | - | 12, 13 | 1319 | ||
AF077040.1 | - | 11, 12 | 1096 | |||
AK022931.1 | - | 10, 11 | 1039 | |||
AK301236.1 | - | 11, 12 | 1236 | |||
AK301563.1 | - | 6, 7 | 751 | |||
AK307810.1 | - | 12, 13 | 1211 | |||
AK309382.1 | - | 11, 12 | 1273 | |||
AK314223.1 | - | 12, 13 | 1371 | |||
AY461579.1 | - | 13, 14 | 1481 | |||
BC018113.2 | - | 12, 13 | 1317 | 3916811 | ||
BC065300.1 | - | 12, 13 | 1352 | 6146202 | ||
BC107137.1 | - | 12, 13 | 1312 | 40025968 | ||
BC107138.1 | - | 12, 13 | 1312 | 40025969 | ||
BT007269.1 | - | 12, 13 | 1223 | |||
BX647760.1 | - | 13, 14 | 1346 |