siRNA Details
Gene Symbol: Smarce1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020618.4 | NP_065643.1 | 6 | 1020 | ||
GenBank mRNA | AK018493.1 | - | 6 | 448 | ||
AK076177.1 | - | 6 | 452 | |||
BC034509.1 | - | 6 | 435 | |||
BC047141.1 | - | 6 | 441 | 6397290 | ||
BC061498.1 | - | 6 | 448 | 5251562 | ||
BC065043.1 | - | 6 | 1020 | 5685483 | ||
EU327022.1 | - | 5 | 345 | |||
EU327023.1 | - | 4 | 301 | |||
EU327024.1 | - | 5 | 398 | |||
EU327025.1 | - | 4 | 293 | |||
EU327026.1 | - | 3 | 249 |

More Information
Other Species Targeted

Panther Classification:
