Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021292.2 | NP_067267.2 | 3 | 430 | ||
GenBank mRNA | AJ250841.1 | - | 3 | 410 | ||
AK039759.1 | - | 3 | 400 | |||
AK148262.1 | - | 3 | 405 | |||
BC021392.1 | - | 3 | 419 | 5069005 | ||
BC021464.1 | - | 3 | 419 | 5069004 |