Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RSAD2 SIRNA Assays ›
          Gene Symbol:
          Rsad2
          Gene Name:
          radical S-adenosyl methionine domain containing 2
          Gene Aliases:
          2510004L01Rik, Vig1, cig5
          Chromosome Location:
          Chr.12: 26442743 - 26456478 on Build GRCm38
          UniGene ID:
          Mm.24045
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 181833

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rsad2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021384.4 NP_067359.2 6 1278
          GenBank mRNA AK010906.1 - 6 1257
          AK036373.1 - 6 1258
          AK149900.1 - 6 1259
          AK150069.1 - 6 1260
          AK150347.1 - 6 1135
          AK150406.1 - 6 1258
          AK150425.1 - 6 1260
          AK150525.1 - 6 1259
          AK150536.1 - 6 1261
          AK150616.1 - 6 1258
          AK150684.1 - 6 1260
          AK150731.1 - 6 1260
          AK150767.1 - 6 1260
          AK150783.1 - 6 1260
          AK150788.1 - 6 1260
          AK150812.1 - 6 1260
          AK150835.1 - 6 1259
          AK151140.1 - 6 1260
          AK151185.1 - 6 1260
          AK151191.1 - 6 1260
          AK151206.1 - 6 1260
          AK151286.1 - 6 1258
          AK151308.1 - 6 1260
          AK151331.1 - 7 1695
          AK151339.1 - 6 1258
          AK151389.1 - 6 1260
          AK151410.1 - 6 1259
          AK151412.1 - 6 1258
          AK151414.1 - 6 1259
          AK151476.1 - 6 1260
          AK151502.1 - 6 1260
          AK151538.1 - 6 1260
          AK151767.1 - 6 1260
          AK151770.1 - 6 1258
          AK151825.1 - 6 1258
          AK151833.1 - 6 1258
          AK151846.1 - 6 1258
          AK151906.1 - 6 1260
          AK151909.1 - 6 1258
          AK151940.1 - 6 1258
          AK151954.1 - 6 1260
          AK151971.1 - 6 1260
          AK152050.1 - 6 1258
          AK152065.1 - 6 1260
          AK152075.1 - 6 1260
          AK152235.1 - 6 1258
          AK152239.1 - 6 1260
          AK152241.1 - 6 1260
          AK152310.1 - 6 1258
          AK152354.1 - 6 1258
          AK152493.1 - 6 1260
          AK152600.1 - 6 1258
          AK152601.1 - 6 1260
          AK152631.1 - 6 1258
          AK152644.1 - 6 1260
          AK152664.1 - 6 1260
          AK152701.1 - 6 1258
          AK152796.1 - 6 1258
          AK152861.1 - 6 1258
          AK152880.1 - 6 1258
          AK152895.1 - 6 1260
          AK152934.1 - 6 1260
          AK152962.1 - 6 1258
          AK153003.1 - 6 1260
          AK153024.1 - 6 1260
          AK153062.1 - 6 1262
          AK153077.1 - 6 1258
          AK153078.1 - 6 1258
          AK153232.1 - 6 1260
          AK153234.1 - 6 1260
          AK153306.1 - 6 1258
          AK153318.1 - 6 1260
          AK153324.1 - 6 1260
          AK153350.1 - 7 1695
          AK153435.1 - 6 1136
          AK153453.1 - 6 1224
          AK153556.1 - 6 1260
          AK159147.1 - 6 1258
          BC057868.1 - 6 1251 5327504

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          ossification
          immune system process
          response to virus
          regulation of ossification
          positive regulation of toll-like receptor 7 signaling pathway
          positive regulation of toll-like receptor 9 signaling pathway
          CD4-positive, alpha-beta T cell activation
          CD4-positive, alpha-beta T cell differentiation
          negative regulation of viral genome replication
          innate immune response
          negative regulation of protein secretion
          defense response to virus
          positive regulation of T-helper 2 cell cytokine production
          catalytic activity
          protein binding
          protein self-association
          metal ion binding
          iron-sulfur cluster binding
          4 iron, 4 sulfur cluster binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0