Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HSPA8 SIRNA Assays ›
          Gene Symbol:
          HSPA8
          Gene Name:
          heat shock protein family A (Hsp70) member 8
          Gene Aliases:
          HEL-33, HEL-S-72p, HSC54, HSC70, HSC71, HSP71, HSP73, HSPA10, LAP-1, LAP1, NIP71
          Chromosome Location:
          Chr.11: 123057492 - 123062366 on Build GRCh38
          UniGene ID:
          Hs.180414
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 18201

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: HSPA8
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_006597.5 NP_006588.1 3 609
          NM_153201.3 NP_694881.1 3 609
          XM_011542798.1 XP_011541100.1 3 357
          GenBank mRNA AB034951.1 - 3 373
          AB073886.1 - 2 287
          AF352832.1 - 3 396
          AK096100.1 - 3 335
          AK129885.1 - 3 409
          AK222628.1 - 3 407
          AK222785.1 - 3 407
          AK300404.1 - 3 483
          AK310467.1 - 3 481
          BC007276.1 - 2 157
          BC016179.1 - 3 390 3920744
          BC016660.1 - 3 390 3923981
          BC019816.2 - 3 390 2899894
          DC395565.1 - 3 542
          EU668349.1 - 3 390
          FJ224294.1 - 3 410

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          mRNA splicing, via spliceosome
          transcription, DNA-templated
          protein folding
          protein targeting to lysosome
          response to unfolded protein
          neurotransmitter secretion
          cellular response to starvation
          viral process
          regulation of protein stability
          protein refolding
          regulation of protein complex assembly
          regulation of mRNA stability
          negative regulation of transcription, DNA-templated
          ATP metabolic process
          positive regulation of mRNA splicing, via spliceosome
          chaperone mediated protein folding requiring cofactor
          regulation of cell cycle
          regulation of protein complex stability
          chaperone-mediated autophagy
          late endosomal microautophagy
          protein targeting to lysosome involved in chaperone-mediated autophagy
          chaperone-mediated protein transport involved in chaperone-mediated autophagy
          clathrin coat disassembly
          cell-cell adhesion
          regulation of cellular response to heat
          negative regulation of fibril organization
          regulation of protein import
          chaperone-mediated autophagy translocation complex disassembly
          G-protein coupled receptor binding
          phosphatidylserine binding
          protein binding
          ATP binding
          ATPase activity
          enzyme binding
          MHC class II protein complex binding
          heat shock protein binding
          ubiquitin protein ligase binding
          ATPase activity, coupled
          poly(A) RNA binding
          unfolded protein binding
          C3HC4-type RING finger domain binding
          cadherin binding involved in cell-cell adhesion

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-9c8cf:80/100.66.75.150:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0