Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sirt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122765.1 | NP_001116237.1 | 8 | 622 | ||
NM_001122766.1 | NP_001116238.1 | 6 | 459 | |||
NM_022432.4 | NP_071877.3 | 9 | 669 | |||
GenBank mRNA | AB221570.1 | - | 9 | 574 | ||
AF299337.1 | - | 9 | 654 | |||
AK014042.1 | - | 8 | 593 | |||
AK144431.1 | - | 8 | 566 | |||
AK146412.1 | - | 8 | 602 | |||
AK183534.1 | - | 1 | 43 | |||
BC021439.1 | - | 9 | 673 | 5039278 | ||
KF032392.1 | - | 9 | 611 |