Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022654.1 | NP_073145.1 | 7, 8 | 1583 | ||
GenBank mRNA | AF274973.1 | - | 7, 8 | 1583 | ||
AK004702.1 | - | 8, 9 | 1526 | |||
AK087880.1 | - | 1803 | ||||
AK156863.1 | - | 9, 10 | 1571 | |||
AK183598.1 | - | 3, 4 | 330 | |||
BC056987.1 | - | 7, 8 | 1934 | |||
BC089373.1 | - | 7, 8 | 1555 | |||
BC099686.1 | - | 7, 8 | 1921 | |||
BC145314.1 | - | 7, 8 | 1587 | |||
BC145315.1 | - | 7, 8 | 1548 | |||
BC145857.1 | - | 7, 8 | 1548 | 40131160 |