Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025343.5 | NP_079619.4 | 3 | 419 | ||
GenBank mRNA | AK007323.1 | - | 3 | 367 | ||
AK075603.1 | - | 3 | 402 | |||
AK169025.1 | - | 3 | 412 | |||
BC036150.1 | - | 2 | 339 | 4977394 | ||
BY740212.1 | - | 3 | 412 |