Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025382.6 | NP_079658.2 | 6 | 896 | ||
GenBank mRNA | AK038972.1 | - | 7 | 1058 | ||
AK138874.1 | - | 1 | 544 | |||
AY364165.1 | - | 6 | 681 | |||
AY846873.1 | - | 6 | 897 | |||
AY846874.1 | - | 6 | 897 | |||
BC019186.1 | - | 6 | 865 | |||
BC042733.2 | - | 6 | 836 | |||
BC043306.1 | - | 3 | 328 | |||
BC044802.1 | - | 6 | 836 | |||
BC054355.1 | - | 7 | 809 | |||
BC054769.1 | - | 6 | 789 | 6415086 |