Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rmdn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025476.6 | NP_079752.3 | 5, 6 | 717 | ||
GenBank mRNA | AK002442.1 | - | 5, 6 | 600 | ||
AK009059.1 | - | 5, 6 | 575 | |||
AK010421.2 | - | 5, 6 | 594 | |||
AK029446.1 | - | 5, 6 | 577 | |||
AK160266.1 | - | 5, 6 | 598 | |||
BC025158.1 | - | 5, 6 | 590 | 4023319 | ||
BC096046.1 | - | 5, 6 | 694 | 1397855 |