Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DHRS7 SIRNA Assays ›
Gene Symbol:
Dhrs7
Gene Name:
dehydrogenase/reductase (SDR family) member 7
Gene Aliases:
2310016E22Rik, 5730564L20Rik, AW061210, Retdsr4, Retsdr4
Chromosome Location:
Chr.12: 72650353 - 72664828 on Build GRCm38
UniGene ID:
Mm.289653
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
182976
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dhrs7
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_025522.5
NP_079798.2
7
1098
GenBank mRNA
AK009385.1
-
7
1077
AK014100.1
-
7
1053
BC016189.1
-
7
1064
4482814
Back To Top
More Information
Panther Classification:
Molecular Function -
dehydrogenase
oxidoreductase
metabolite interconversion enzyme
Gene Ontology Categories:
Function(s)
Process(es)
biological_process
metabolic process
oxidation-reduction process
oxidoreductase activity
pinocarveol dehydrogenase activity
chloral hydrate dehydrogenase activity
hydroxymethylmethylsilanediol oxidase activity
1-phenylethanol dehydrogenase activity
myrtenol dehydrogenase activity
cis-1,2-dihydroxy-1,2-dihydro-8-carboxynaphthalene dehydrogenase activity
3-hydroxy-4-methyloctanoyl-CoA dehydrogenase activity
2-hydroxy-4-isopropenylcyclohexane-1-carboxyl-CoA dehydrogenase activity
cis-9,10-dihydroanthracene-9,10-diol dehydrogenase activity
citronellol dehydrogenase activity
naphthyl-2-hydroxymethyl-succinyl-CoA dehydrogenase activity
2,4,4-trimethyl-1-pentanol dehydrogenase activity
2,4,4-trimethyl-3-hydroxypentanoyl-CoA dehydrogenase activity
1-hydroxy-4,4-dimethylpentan-3-one dehydrogenase activity
endosulfan diol dehydrogenase activity
endosulfan hydroxyether dehydrogenase activity
3-hydroxy-2-methylhexanoyl-CoA dehydrogenase activity
3-hydroxy-2,6-dimethyl-5-methylene-heptanoyl-CoA dehydrogenase activity
versicolorin reductase activity
ketoreductase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-zcc2s:80/100.66.79.68:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0