Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025531.2 | NP_079807.1 | 3 | 300 | ||
GenBank mRNA | AK003043.1 | - | 3 | 274 | ||
AK008542.1 | - | 3 | 260 | |||
AK013197.1 | - | 3 | 267 | |||
AK150668.1 | - | 3 | 276 | |||
BC004775.1 | - | 2 | 98 | |||
BC026968.1 | - | 3 | 300 | 4235395 |