Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025675.4 | NP_079951.1 | 5 | 491 | ||
GenBank mRNA | AK006965.1 | - | 5 | 449 | ||
AK007021.1 | - | 5 | 494 | |||
AK017595.1 | - | 5 | 447 | |||
AK036021.1 | - | 5 | 527 | |||
AK082013.1 | - | 5 | 528 | |||
AK088160.1 | - | 5 | 493 | |||
AK145789.1 | - | 5 | 451 | |||
BC022995.1 | - | 5 | 516 | 5363234 |