Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pacrgl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025755.3 | NP_080031.1 | 3 | 311 | ||
GenBank mRNA | AK014276.1 | - | 3 | 324 | ||
AK016966.1 | - | 3 | 360 | |||
AK018101.1 | - | 3 | 321 | |||
AK032498.1 | - | 2 | 2059 | |||
BC018481.1 | - | 3 | 311 | 3482169 |