Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Efcab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025769.3 | NP_080045.1 | 2 | 257 | ||
GenBank mRNA | AK015426.1 | - | 239 | |||
AK015866.1 | - | 2 | 277 | |||
AK017267.1 | - | 2 | 258 | |||
AK030381.1 | - | 2 | 267 | |||
AK038735.1 | - | 2 | 267 | |||
AK042748.1 | - | 2 | 299 | |||
AK043519.1 | - | 2 | 229 | |||
AK077722.1 | - | 2 | 260 | |||
BC085167.1 | - | 2 | 149 | 30538702 |