Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159518.1 | NP_001152990.1 | 6 | 1070 | ||
NM_008048.3 | NP_032074.3 | 5 | 977 | |||
GenBank mRNA | AB012886.1 | - | 5 | 978 | ||
AK002277.1 | - | 5 | 973 | |||
AK148749.1 | - | 5 | 974 | |||
AK156315.1 | - | 5 | 979 | |||
BC047202.1 | - | 5 | 889 | |||
BC092538.1 | - | 5 | 973 | 30284911 | ||
L75822.2 | - | 5 | 955 |