Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SEPT9 SIRNA Assays ›
Gene Symbol:
SEPT9
Gene Name:
septin 9
Gene Aliases:
AF17q25, MSF, MSF1, NAPB, PNUTL4, SINT1, SeptD1
Chromosome Location:
Chr.17: 77281410 - 77500596 on Build GRCh38
UniGene ID:
Hs.440932
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
18412
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SEPT9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001113491.1
NP_001106963.1
12
2674
NM_001113492.1
NP_001106964.1
12
2849
NM_001113493.1
NP_001106965.1
11
2835
NM_001113494.1
NP_001106966.1
11
2788
NM_001113495.1
NP_001106967.1
10
2488
NM_001113496.1
NP_001106968.1
10
2083
NM_001293695.1
NP_001280624.1
11
2617
NM_001293696.1
NP_001280625.1
10
1899
NM_001293697.1
NP_001280626.1
10
2112
NM_001293698.1
NP_001280627.1
10
2198
NM_006640.4
NP_006631.2
11
3306
XM_005256962.1
XP_005257019.1
10
1956
XM_006721643.2
XP_006721706.1
13
2777
XM_006721644.1
XP_006721707.1
10
1999
XM_011524204.1
XP_011522506.1
13
2874
XM_011524206.1
XP_011522508.1
11
2594
XM_011524207.1
XP_011522509.1
11
2607
XM_011524208.2
XP_011522510.1
10
1865
XM_017024031.1
XP_016879520.1
11
2206
XM_017024032.1
XP_016879521.1
10
2205
GenBank mRNA
AB023208.1
-
11
2789
AF142408.1
-
11
2747
AF142569.1
-
1
701
AF189712.2
-
13
2802
AF189713.3
-
12
2645
AJ312319.1
-
11
2836
AJ312320.1
-
10
1878
AK022493.1
-
6
763
AL080131.1
-
7
1494
BC021192.2
-
11
2530
3346534
BC054004.1
-
13
2505
6146448
BQ279153.1
-
415
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
cell cycle
protein heterooligomerization
cell division
cell-cell adhesion
positive regulation of nonmotile primary cilium assembly
GTPase activity
protein binding
GTP binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0