Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Timm13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013895.4 | NP_038923.1 | 3 | 746 | ||
GenBank mRNA | AK009041.1 | - | 3 | 540 | ||
AK009152.1 | - | 3 | 556 | |||
AK013230.1 | - | 3 | 544 | |||
AK088984.1 | - | 3 | 556 | |||
AK213641.1 | - | 1 | 156 | |||
BC011436.1 | - | 3 | 559 | 2812494 |