Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015767.3 | NP_056582.1 | 5 | 927 | ||
GenBank mRNA | AK004882.1 | - | 5 | 908 | ||
AK050374.1 | - | 5 | 928 | |||
BC026882.1 | - | 5 | 814 | |||
BC043705.1 | - | 1 | 482 |