Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015826.5 | NP_056641.2 | 3 | 694 | ||
GenBank mRNA | AB221653.1 | - | 3 | 546 | ||
AF192561.1 | - | 3 | 237 | |||
AF202778.1 | - | 3 | 681 | |||
AL133300.1 | - | 3 | 697 | |||
AY169786.1 | - | 3 | 697 | |||
AY169787.1 | - | 3 | 697 | |||
AY943925.1 | - | 3 | 236 | |||
BC125474.1 | - | 3 | 657 | 40129989 | ||
DQ530630.1 | - | 3 | 693 |