Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015826.5 | NP_056641.2 | 5 | 1185 | ||
GenBank mRNA | AB221653.1 | - | 5 | 1037 | ||
AF202778.1 | - | 5 | 1172 | |||
AL133300.1 | - | 5 | 1188 | |||
AY169785.1 | - | 4 | 904 | |||
AY169786.1 | - | 5 | 1188 | |||
AY169787.1 | - | 5 | 1188 | |||
AY943925.1 | - | 5 | 727 | |||
BC125474.1 | - | 5 | 1148 | 40129989 |