Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Copg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017477.2 | NP_059505.1 | 17 | 1793 | ||
GenBank mRNA | AF187079.1 | - | 17 | 1792 | ||
AK049417.1 | - | 17 | 1744 | |||
AK077461.1 | - | 17 | 1740 | |||
AK147895.1 | - | 17 | 1806 | |||
AK151284.1 | - | 17 | 1798 | |||
AK151816.1 | - | 17 | 1798 | |||
AK153142.1 | - | 17 | 1798 | |||
AK164280.1 | - | 17 | 1763 | |||
AK169256.1 | - | 17 | 1838 | |||
AK171950.1 | - | 17 | 1772 | |||
AK172519.1 | - | 17 | 1809 | |||
BC024896.1 | - | 17 | 1761 | 4189311 | ||
BC060744.1 | - | 3 | 2885 |