Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eif3d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018749.2 | NP_061219.2 | 14 | 1576 | ||
GenBank mRNA | AB012580.1 | - | 14 | 1517 | ||
AK045228.1 | - | 14 | 1518 | |||
AK049767.1 | - | 14 | 1515 | |||
AK145689.1 | - | 14 | 1531 | |||
AK168256.1 | - | 14 | 1566 | |||
AK205598.1 | - | 1 | 57 | |||
BC089020.1 | - | 14 | 1510 | 30551991 |