siRNA Details
Gene Symbol: Camkk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018883.2 | NP_061371.2 | 5, 6 | 667 | ||
GenBank mRNA | AF117384.1 | - | 5, 6 | 622 | ||
AK144483.1 | - | 5, 6 | 668 | |||
BC017529.1 | - | 5, 6 | 632 | 4924656 |

More Information
Other Species Targeted

Panther Classification:
