Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mefv | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161790.1 | NP_001155262.1 | 2 | 696 | ||
NM_001161791.1 | NP_001155263.1 | 2 | 696 | |||
NM_019453.2 | NP_062326.2 | 2 | 696 | |||
GenBank mRNA | AF143409.1 | - | 2 | 669 | ||
BC108993.1 | - | 2 | 657 | 40047838 | ||
BC108994.1 | - | 2 | 657 | 40047839 |