Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Crtam | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281954.1 | NP_001268883.1 | 9 | 1041 | ||
NM_019465.4 | NP_062338.3 | 9 | 1038 | |||
GenBank mRNA | AF001104.1 | - | 9 | 984 | ||
AK005719.1 | - | 7 | 764 | |||
AK163744.1 | - | 4 | 439 | |||
BC117715.1 | - | 9 | 986 | |||
BC117716.1 | - | 9 | 987 | 40088099 |