Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il17b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019508.1 | NP_062381.1 | 3 | 665 | ||
GenBank mRNA | AF184970.1 | - | 3 | 665 | ||
AF218726.1 | - | 3 | 665 | |||
AF250308.1 | - | 3 | 662 | |||
AK003506.1 | - | 3 | 723 | |||
BC002271.1 | - | 3 | 665 | 3496106 |