Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLLT11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006818.3 | NP_006809.1 | 2 | 978 | ||
GenBank mRNA | AK056089.1 | - | 2 | 973 | ||
AK057755.1 | - | 2 | 248 | |||
BC006471.2 | - | 1 | 100 | 2823316 | ||
BC007079.1 | - | 2 | 242 | |||
BC008445.1 | - | 2 | 242 | 4103485 | ||
BC009624.1 | - | 2 | 258 | 3878959 | ||
BC019253.1 | - | 2 | 218 | 3502660 | ||
BC021703.1 | - | 2 | 316 | 4108071 | ||
BC022448.1 | - | 2 | 1250 | 4815214 | ||
BC032996.1 | - | 2 | 275 | |||
BT006799.1 | - | 1 | 93 | |||
CR456879.1 | - | 1 | 93 | |||
CR542150.1 | - | 1 | 93 | |||
U16954.1 | - | 2 | 448 |