Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Styx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019637.3 | NP_062611.2 | 11 | 1210 | ||
GenBank mRNA | AK030122.1 | - | 11 | 1210 | ||
AK031186.1 | - | 1 | 272 | |||
AK042741.1 | - | 12 | 1265 | |||
AK046970.1 | - | 11 | 1187 | |||
AK154136.1 | - | 11 | 1305 | |||
AK166043.1 | - | 11 | 1193 |