Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Diap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019670.1 | NP_062644.1 | 11 | 1079 | ||
GenBank mRNA | AF094519.1 | - | 11 | 1079 | ||
AK137073.1 | - | 10 | 1168 | |||
AK138694.1 | - | 11 | 1109 | |||
AK161737.1 | - | 10 | 1097 | |||
AK166882.1 | - | 11 | 1135 | |||
AK220493.1 | - | 11 | 1143 | |||
BC003250.1 | - | 10 | 1113 | |||
BC028920.1 | - | 4 | 380 | 4008236 | ||
BC085191.1 | - | 11 | 1189 | 30693873 | ||
BC086779.1 | - | 11 | 1154 |