Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019793.3 | NP_062767.3 | 2 | 326 | ||
GenBank mRNA | AB009021.1 | - | 2 | 108 | ||
AB072450.1 | - | 2 | 136 | |||
AF133427.1 | - | 2 | 120 | |||
AF242591.1 | - | 2 | 319 | |||
AK006798.1 | - | 2 | 202 | |||
AK053325.1 | - | 2 | 177 | |||
AK089494.1 | - | 158 | ||||
AK141951.1 | - | 2 | 184 | |||
AK146130.1 | - | 2 | 314 | |||
AK152175.1 | - | 2 | 211 | |||
AK154919.1 | - | 158 | ||||
AK159324.1 | - | 2 | 297 | |||
AK159418.1 | - | 2 | 270 | |||
AK159621.1 | - | 2 | 281 | |||
AK159914.1 | - | 2 | 259 | |||
AK168231.1 | - | 2 | 314 | |||
AK169068.1 | - | 2 | 297 | |||
AK212256.1 | - | 1 | 32 | |||
AK213393.1 | - | 1 | 32 | |||
AK213564.1 | - | 1 | 32 | |||
AK218357.1 | - | 1 | 32 | |||
BC043072.1 | - | 2 | 304 | 6404165 |