Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctsf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019861.1 | NP_063914.1 | 5 | 659 | ||
GenBank mRNA | AF136280.1 | - | 5 | 560 | ||
AF197480.1 | - | 5 | 604 | |||
AJ131851.1 | - | 5 | 536 | |||
AK017474.1 | - | 5 | 545 | |||
AK075862.1 | - | 5 | 552 | |||
AK165416.1 | - | 4 | 719 | |||
BC004054.1 | - | 2 | 130 | |||
BC051131.1 | - | 4 | 778 | |||
BC058758.1 | - | 5 | 550 | 6492834 |