Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture and Transfection
    • Chemicals
    • Chromatography
    • Electron Microscopes
    • Lab Plasticware and Supplies
    • Lab Centrifuges
    • Lab Solutions
    • Mass Spectrometers
    • Next Generation Sequencers
    • See all product categories
  • Applications
    • Brands
    • Industrial and Applied Sciences
    • Food and Beverage
    • Forensics
    • Lab Solutions
    • Life Sciences
    • Pharma and Biopharma
    • Biotechnology
    • Clinical and Diagnostics
    • Digital Solutions
    • See all applications
  • Services
    • Lab Instrument and Equipment Services
    • Custom Services
    • Training Services
    • Financial and Leasing Services
    • Enterprise Level Lab Informatics
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Cell Biology Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • Contact Us
    • Product Documentation
    • Knowledge Base and Product FAQs
    • Learning Centers
    • Supply Center
    • eProcurement Solutions
    • Lab Instrument and Equipment Support
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Special Offers
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTCH1 SIRNA Assays ›
          Gene Symbol:
          Mtch1
          Gene Name:
          mitochondrial carrier homolog 1 (C. elegans)
          Gene Aliases:
          2310034O17Rik, AI255158, AU018396, C77849, PSAP
          Chromosome Location:
          Chr.17: 29332076 - 29347927 on Build GRCm38
          UniGene ID:
          Mm.285322
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 185724

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mtch1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_019880.3 NP_063933.1 12 1889
          GenBank mRNA AF176007.2 - 12 1839
          AF192558.2 - 12 1890
          AK009623.1 - 12 1776
          AK154010.1 - 12 1694
          AK179013.1 - 1 269
          AK182222.1 - 1 269
          AK189748.1 - 2 235
          AK191792.1 - 1 269
          AK191954.1 - 1 269
          AK192009.1 - 269
          AK194435.1 - 1 270
          AK194668.1 - 1 271
          AK195206.1 - 2 216
          AK196523.1 - 1 295
          AK196870.1 - 1 269
          AK196930.1 - 1 269
          AK197105.1 - 1 269
          AK197332.1 - 1 269
          AK197964.1 - 1 269
          AK198074.1 - 1 269
          AK198564.1 - 1 269
          AK198904.1 - 2 216
          AK199020.1 - 1 349
          AK199094.1 - 1 269
          AK199530.1 - 1 276
          AK200252.1 - 1 357
          AK201056.1 - 1 269
          AK201212.1 - 1 269
          AK201278.1 - 1 254
          AK203000.1 - 1 268
          AK205205.1 - 1 339
          AK205452.1 - 1 269
          AK205989.1 - 1 269
          AK206098.1 - 1 269
          AK206512.1 - 1 269
          AK206937.1 - 1 269
          AK207126.1 - 1 269
          AK207230.1 - 1 269
          AK207280.1 - 1 269
          AK208701.1 - 1 269
          AK208828.1 - 1 270
          AK209213.1 - 1 269
          AK209427.1 - 1 269
          AK209530.1 - 1 269
          AK209577.1 - 1 269
          AK209789.1 - 1 269
          AK209850.1 - 2 216
          AK209981.1 - 1 269
          AK211639.1 - 1 339
          AK211746.1 - 1 269
          AK212498.1 - 1 297
          AK213297.1 - 1 269
          AK214524.1 - 1 269
          AK215059.1 - 1 269
          AK215161.1 - 1 268
          AK215190.1 - 1 269
          AK216129.1 - 1 269
          AK216414.1 - 2 216
          AK217409.1 - 1 269
          AK218222.1 - 1 270
          AK218238.1 - 1 269
          AK220070.1 - 1 269
          BC023135.1 - 12 1732 3669288
          BC026436.1 - 8 1123
          BC027274.1 - 12 1687 3156729
          BC029723.1 - 1 2254

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          mitochondrial carrier protein

          Gene Ontology Categories:

          Function(s)

          transport
          apoptotic process
          activation of cysteine-type endopeptidase activity involved in apoptotic process

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Quick Order
          • eProcurement
          • Supply Center
          • Order Status
          • Chemicals
          • India Mobile App
          • Government eMarketplace
          Support Plus Icon Minus Icon
          • Order Support
          • Training
          • Contact Us
          • Report a Site Issue
          • Instrument Management
          Resources Plus Icon Minus Icon
          • Product Selection Guides
          • Mobile & Desktop Apps
          • Webinars
          • Blog 
          • Social Media
          • New Products
          • Promotions
          • Shared Lists
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          India flag icon
          India

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0