Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Instrument Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • Our Instagram
      Our Instagram
    • Our Facebook
      Our Facebook
    • Blog Behind the Bench
      Blog Behind the Bench
    • Customer Experience Center (CEC)
      Customer Experience Center (CEC)
    • Ecommerce Exclusives
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MTOR SIRNA Assays ›
          Gene Symbol:
          Mtor
          Gene Name:
          mechanistic target of rapamycin (serine/threonine kinase)
          Gene Aliases:
          2610315D21Rik, AI327068, FRAP, FRAP2, Frap1, RAFT1, RAPT1, flat
          Chromosome Location:
          Chr.4: 148448582 - 148557685 on Build GRCm38
          UniGene ID:
          Mm.21158
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 185896

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Mtor
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_020009.2 NP_064393.2 8 1276
          GenBank mRNA AF152838.1 - 8 1251
          AK143381.1 - 5 687
          AK172584.1 - 8 1267
          BC112904.1 - 8 1234 6465126

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase protein modifying enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          double-strand break repair via homologous recombination
          positive regulation of protein phosphorylation
          positive regulation of endothelial cell proliferation
          regulation of glycogen biosynthetic process
          regulation of carbohydrate metabolic process
          protein phosphorylation
          response to stress
          germ cell development
          negative regulation of autophagy
          positive regulation of lamellipodium assembly
          positive regulation of gene expression
          positive regulation of myotube differentiation
          cell growth
          negative regulation of macroautophagy
          phosphorylation
          peptidyl-serine phosphorylation
          peptidyl-threonine phosphorylation
          cell projection organization
          positive regulation of actin filament polymerization
          ruffle organization
          regulation of myelination
          cellular response to nutrient levels
          TOR signaling
          regulation of fatty acid beta-oxidation
          regulation of response to food
          response to insulin
          regulation of actin cytoskeleton organization
          regulation of GTPase activity
          response to amino acid
          regulation of carbohydrate utilization
          regulation of osteoclast differentiation
          positive regulation of translation
          negative regulation of cell size
          regulation of protein kinase activity
          positive regulation of transcription from RNA polymerase III promoter
          protein autophosphorylation
          positive regulation of lipid biosynthetic process
          positive regulation of peptidyl-tyrosine phosphorylation
          positive regulation of stress fiber assembly
          negative regulation of NFAT protein import into nucleus
          positive regulation of protein kinase B signaling
          cellular response to hypoxia
          nucleotide binding
          RNA polymerase III type 1 promoter DNA binding
          RNA polymerase III type 2 promoter DNA binding
          RNA polymerase III type 3 promoter DNA binding
          TFIIIC-class transcription factor binding
          protein serine/threonine kinase activity
          protein binding
          ATP binding
          drug binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          phosphotransferase activity, alcohol group as acceptor
          protein domain specific binding
          ribosome binding
          protein dimerization activity
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Find Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Brasil flag icon
          Brasil

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0