Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006773.3 | NP_006764.3 | 3 | 507 | ||
GenBank mRNA | AK001467.1 | - | 3 | 479 | ||
BC001238.1 | - | 3 | 436 | 3452935 | ||
BC003360.1 | - | 3 | 466 | 2900252 | ||
BC024739.1 | - | 3 | 481 | 4995871 | ||
CR457060.1 | - | 2 | 228 | |||
X98743.1 | - | 2 | 245 |