Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhrs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037938.2 | NP_001033027.2 | 4 | 572 | ||
NM_030686.2 | NP_109611.1 | 4 | 572 | |||
GenBank mRNA | AB045132.1 | - | 4 | 484 | ||
AK018788.1 | - | 4 | 455 | |||
AK146988.1 | - | 4 | 452 | |||
BC003484.1 | - | 4 | 407 | 2811468 | ||
BC054361.1 | - | 4 | 533 | 5008299 |