Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Trpm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138301.2 | NP_612174.2 | 3, 4 | 487 | ||
GenBank mRNA | AB166747.1 | - | 3, 4 | 469 | ||
AJ344343.1 | - | 1, 2 | 615 | |||
AJ878415.1 | - | 1, 2 | 170 | |||
AK039405.1 | - | 3, 4 | 414 | |||
BC141391.1 | - | 1, 2 | 200 | 9056004 | ||
CJ108999.1 | - | 3, 4 | 488 |