Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCNI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006835.2 | NP_006826.1 | 4, 5 | 861 | ||
GenBank mRNA | AB209766.1 | - | 4, 5 | 863 | ||
AF135162.1 | - | 4, 5 | 516 | |||
AK222931.1 | - | 3, 4 | 313 | |||
AK301111.1 | - | 3, 4 | 722 | |||
AK312632.1 | - | 6, 7 | 731 | |||
BC000420.2 | - | 4, 5 | 860 | 2964432 | ||
BC004975.1 | - | 4, 5 | 860 | 2957878 | ||
BT020019.1 | - | 3, 4 | 317 | |||
CR541783.1 | - | 3, 4 | 317 | |||
D50310.1 | - | 3, 4 | 317 |