Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SIRT6 SIRNA Assays ›
          Gene Symbol:
          Sirt6
          Gene Name:
          sirtuin 6
          Gene Aliases:
          2810449N18Rik, AI043036, Sir2l6
          Chromosome Location:
          Chr.10: 81621786 - 81627792 on Build GRCm38
          UniGene ID:
          Mm.131825
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 186534

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sirt6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_181586.3 NP_853617.1 8 1189
          GenBank mRNA AK013316.1 - 8 1137
          BC052763.1 - 8 1139 6513375

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          post-embryonic cardiac muscle cell growth involved in heart morphogenesis
          protein ADP-ribosylation
          regulation of double-strand break repair via homologous recombination
          histone H3-K9 modification
          histone H3-K9 deacetylation
          chromatin binding
          NAD(P)+-protein-arginine ADP-ribosyltransferase activity
          histone deacetylase activity
          protein-N-terminal asparagine amidohydrolase activity
          UDP-3-O-[3-hydroxymyristoyl] N-acetylglucosamine deacetylase activity
          hydrolase activity
          iprodione amidohydrolase activity
          (3,5-dichlorophenylurea)acetate amidohydrolase activity
          protein deacetylase activity
          4'-(2-hydroxyisopropyl)phenylurea amidohydrolase activity
          didemethylisoproturon amidohydrolase activity
          N-isopropylacetanilide amidohydrolase activity
          N-cyclohexylformamide amidohydrolase activity
          isonicotinic acid hydrazide hydrolase activity
          cis-aconitamide amidase activity
          gamma-N-formylaminovinylacetate hydrolase activity
          N2-acetyl-L-lysine deacetylase activity
          O-succinylbenzoate synthase activity
          indoleacetamide hydrolase activity
          N-acetylcitrulline deacetylase activity
          metal ion binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          N-acetylgalactosamine-6-phosphate deacetylase activity
          diacetylchitobiose deacetylase activity
          chitooligosaccharide deacetylase activity
          NAD+ binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0