Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TERT SIRNA Assays ›
Gene Symbol:
Tert
Gene Name:
telomerase reverse transcriptase
Gene Aliases:
EST2, TCS1, TP2, TR, TRT
Chromosome Location:
Chr.13: 73626797 - 73649843 on Build GRCm38
UniGene ID:
Mm.10109
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
186677
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tert
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_009354.1
NP_033380.1
2
649
GenBank mRNA
AF051911.1
-
2
649
AF073311.1
-
2
620
AK133783.1
-
2
740
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
transcription, RNA-templated
RNA-dependent DNA biosynthetic process
telomere maintenance via telomerase
mitochondrion organization
negative regulation of gene expression
DNA strand elongation
positive regulation of Wnt signaling pathway
production of siRNA involved in RNA interference
RNA biosynthetic process
positive regulation of hair cycle
positive regulation of nitric-oxide synthase activity
cellular response to hypoxia
DNA biosynthetic process
replicative senescence
positive regulation of pri-miRNA transcription from RNA polymerase II promoter
negative regulation of production of siRNA involved in RNA interference
positive regulation of stem cell proliferation
negative regulation of cellular senescence
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
tRNA binding
transcription coactivator binding
DNA binding
telomerase activity
telomerase RNA reverse transcriptase activity
RNA binding
RNA-directed DNA polymerase activity
RNA-directed RNA polymerase activity
protein binding
transferase activity
nucleotidyltransferase activity
telomeric DNA binding
protein homodimerization activity
metal ion binding
telomerase RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0