Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ppp2r5d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009358.3 | NP_033384.2 | 3 | 336 | ||
GenBank mRNA | AB053464.1 | - | 3 | 281 | ||
AF316616.2 | - | 3 | 193 | |||
AK154050.1 | - | 3 | 312 | |||
AK160587.1 | - | 3 | 277 | |||
AY338234.1 | - | 3 | 266 | |||
BC010716.1 | - | 3 | 249 | 2655861 | ||
CJ042073.1 | - | 3 | 337 |