Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYOT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135940.1 | NP_001129412.1 | 3 | 289 | ||
NM_001300911.1 | NP_001287840.1 | 4 | 420 | |||
NM_006790.2 | NP_006781.1 | 3 | 771 | |||
XM_017010060.1 | XP_016865549.1 | 4 | 994 | |||
XM_017010061.1 | XP_016865550.1 | 2 | 427 | |||
XM_017010062.1 | XP_016865551.1 | 3 | 863 | |||
GenBank mRNA | AF133820.2 | - | 3 | 756 | ||
AF144477.1 | - | 3 | 742 | |||
AK300076.1 | - | 4 | 400 | |||
AK300088.1 | - | 3 | 287 | |||
AK307620.1 | - | 4 | 634 | |||
AK312733.1 | - | 3 | 751 | |||
BC005376.1 | - | 3 | 751 | 3934230 | ||
KU178476.1 | - | 2 | 462 |