Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Uty | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009484.2 | NP_033510.2 | 9 | 854 | ||
GenBank mRNA | AF057367.1 | - | 9 | 834 | ||
AK043154.1 | - | 7 | 674 | |||
AK142344.1 | - | 11 | 942 | |||
AK142512.1 | - | 9 | 770 | |||
AK155922.1 | - | 10 | 734 | |||
AK162410.1 | - | 8 | 562 | |||
AK220463.1 | - | 10 | 942 | |||
BC053061.1 | - | 9 | 837 |