Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRSS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270452.1 | NP_001257381.1 | 5 | 727 | ||
NM_006799.3 | NP_006790.1 | 5 | 759 | |||
NM_144956.2 | NP_659205.1 | 5 | 753 | |||
NM_144957.2 | NP_659206.1 | 5 | 759 | |||
NR_073012.1 | 5 | 677 | ||||
XM_017022875.1 | XP_016878364.1 | 5 | 677 | |||
XM_017022876.1 | XP_016878365.1 | 5 | 653 | |||
XM_017022877.1 | XP_016878366.1 | 5 | 636 | |||
XM_017022878.1 | XP_016878367.1 | 5 | 653 | |||
GenBank mRNA | AB031329.1 | - | 5 | 662 | ||
AB031330.1 | - | 5 | 656 | |||
AB031331.1 | - | 5 | 742 | |||
AF058300.1 | - | 5 | 671 | |||
AY359034.1 | - | 5 | 672 | |||
BC074999.2 | - | 5 | 695 | 30915610 | ||
BC075000.2 | - | 5 | 695 | 30915398 | ||
BG717268.1 | - | 5 | 564 | |||
BI334059.1 | - | 3 | 478 | |||
BI827608.1 | - | 5 | 598 |