Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wee1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009516.3 | NP_033542.2 | 3, 4 | 1134 | ||
GenBank mRNA | AK011212.1 | - | 3, 4 | 910 | ||
AK039572.1 | - | 3, 4 | 450 | |||
BC006852.1 | - | 3, 4 | 1153 | 3156953 | ||
D30743.1 | - | 3, 4 | 1135 |