Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009541.2 | NP_033567.2 | 14 | 2084 | ||
GenBank mRNA | AK051971.1 | - | 13 | 2155 | ||
AK134251.1 | - | 14 | 2044 | |||
BC060055.1 | - | 11 | 2354 | |||
BC145312.1 | - | 13 | 1957 | 9053867 | ||
BC145313.1 | - | 12 | 2084 | |||
BC145821.1 | - | 13 | 1957 | 40131124 | ||
U14556.1 | - | 14 | 2082 | |||
U22396.1 | - | 14 | 2072 |