Search
Search

Zfp46| Gene Symbol: Zfp46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009557.3 | NP_033583.2 | 3 | 2536 | ||
| GenBank mRNA | AK051031.1 | - | 3 | 2509 | ||
| AK053652.1 | - | 3 | 2541 | |||
| AK136405.1 | - | 3 | 2445 | |||
| AK148255.1 | - | 3 | 2487 | |||
| AK173240.1 | - | 3 | 2524 | |||
| BC092398.1 | - | 1 | 1846 | |||
| M98502.1 | - | 1 | 2059 | |||