Search
Search

Tekt1| Gene Symbol: Tekt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282006.1 | NP_001268935.1 | 5 | 690 | ||
| NM_001282007.1 | NP_001268936.1 | 5 | 639 | |||
| NM_011569.3 | NP_035699.2 | 5 | 642 | |||
| GenBank mRNA | AF081947.1 | - | 5 | 633 | ||
| AK005801.1 | - | 5 | 691 | |||
| AK076858.1 | - | 5 | 621 | |||
| BC050967.1 | - | 5 | 640 | |||
| BC098181.1 | - | 5 | 600 | 6312556 | ||