Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011669.3 | NP_035799.1 | 2 | 410 | ||
GenBank mRNA | AF441835.1 | - | 1 | 71 | ||
AK006739.1 | - | 2 | 460 | |||
AK035629.1 | - | 2 | 251 | |||
AK167153.1 | - | 2 | 250 | |||
BC068136.1 | - | 2 | 410 | 30363916 |